Catharanthine tartrate (aus Catharanthus roseus) ≥90% (Trockenmasse)

Lieferant: ENZO LIFE SCIENCES

Synonyme: 3,4-Didehydroibogamine-18-carboxylic acid methyl ester tartrate

ALX350101M100
ENZOALX350101M100EA 175 EUR
ENZOALX350101M100
Catharanthine tartrate (aus Catharanthus roseus) ≥90% (Trockenmasse)
Catharanthine tartrate

Starting material for the synthesis of the antitumor drugs vinblastine and vincristine. It is less active as an inhibitor of tubulin self-assembly into microtubules than the latter two compounds.


White to off-white crystalline powder.


Soluble in 100% ethanol.

Formel: C₂₁H₂₄N₂O₂·C₄H₆O₆
Molecular Weight: 336,4 · 150,1 g/mol
Lagertemperatur: Gefrierschrank
CAS-Nummer: 4168-17-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR