Vindoline ≥90% (Trockenmasse)
Lieferant: ENZO LIFE SCIENCES
|
Achtung
|
ALX350102M100
ENZOALX350102M100EA
172
EUR
ENZOALX350102M100
Vindoline ≥90% (Trockenmasse)
Vindoline
Lacks physiological activity itself, but is contained as the pentacyclic moiety in the antineoplastic agents vinblastine and vincristine.
Very hygroscopic and soluble in 100% ethanol and water (30 mg/ml).
Formel:
C₂₅H₃₂N₂O₆ Molecular Weight: 456,5 g/mol Lagertemperatur: Gefrierschrank |
MDL:
MFCD27991347 CAS-Nummer: 2182-14-1 EINECS: 218-558-0 Merck Index: 14,09988 |
Spezifikation Testergebnisse
Purity | ≥90% (Assay on Dried Basis) |
Source | Isolated from Catharanthus roseus |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...