Pseudolaric acid B ≥95%
Lieferant: ENZO LIFE SCIENCES
Synonyme:
Pseudolarix acid B, (2E,4E)-5-((3R,4S,4aS,9aR)-4a-Acetoxy-7-(methoxycarbonyl)-3-methyl-1-oxo-3,4,4a,5,6,9-hexahydro-1H-4,9a-ethanocyclohepta[c]pyran-3-yl)-2-methylpenta-2,4-dienoic acid
Activates PPAR alpha and the phospholipase C (PLC) signalling pathway. Stimulates peroxisomal fatty acyl-CoA oxidase activity. These effects can be blocked by staurosporine (Prod. No. ALX-380-014). Exhibits significant cytotoxic activities against numerous tumor cell lines. Binds tubulin. Inhibits angiogenesis. Reduces HIF-1alpha protein levels. Induces apoptosis. Antifungal.
Isolated from Pseudolarix kaempferi.
Formel:
C₂₃H₂₈O₈ Molecular Weight: 432,5 g/mol |
MDL:
MFCD01746346 CAS-Nummer: 82508-31-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...