Panepoxydone (aus Lentinus crinitus) ≥95% (durch HPLC, MS)

Lieferant: ENZO LIFE SCIENCES

ALX350109MC05
ENZOALX350109MC05EA 634 EUR
ENZOALX350109MC05
Panepoxydone (aus Lentinus crinitus) ≥95% (durch HPLC, MS)
Panepoxydone

Antibiotic. Inhibitor of NF-κB activation by preventing the phosphorylation of IκB protein and thereby interrupts the signalling pathway. Strongly inhibits the expression of several NF-κB dependent pro-inflammatory genes.


  • NF-κB pathway inhibitor


Yellow oil.


Soluble in DMSO, 100% ethanol or methanol.

Formel: C₁₁H₁₄O₄
Molecular Weight: 210.2 g/mol
Lagertemperatur: Gefrierschrank
CAS-Nummer: 31298-54-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR