Sibutramine hydrochloride monohydrate ≥98% (durch HPLC)

Lieferant: ENZO LIFE SCIENCES
Achtung

Synonyme: 1-(4-Chlorophenyl-N,N-dimethyl-α-(2-methylpropyl)cyclobutanemethanamine.HCl.H₂O

ALX350111G001
ENZOALX350111G001EA 103 EUR
ENZOALX350111G001
Sibutramine hydrochloride monohydrate ≥98% (durch HPLC)
Sibutramine hydrochloride monohydrate

Anti-obesity agent.


Prevents resorption of the messenger substances serotonin and noradrenalin in the brain, allowing them to act at higher concentration on nerve cells and increasing the feeling of satiety.


Soluble in methanol or water (2.9 mg/ml at pH 5.2).

Formel: C₁₇H₂₆ClN·HCl·H₂O
Molecular Weight: 334.4 g/mol
Schmelzpunkt: 190…201 °C
Lagertemperatur: Kühlschrank
CAS-Nummer: 125494-59-9
Merck Index: 14,08485

Bestellen

Spezifikation Testergebnisse

Purity ≥98% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR