Neobavaisoflavone ≥97% (durch HPLC)
Lieferant: ENZO LIFE SCIENCES
ALX350146M001
ENZOALX350146M001EA
283
EUR
ENZOALX350146M001
Neobavaisoflavone ≥97% (durch HPLC)
Neobavaisoflavone
Inhibits platelet aggregation. DNA polymerase inhibitor. Shows antifungal activity.
- DNA polymerase inhibitor
Yellow solid.
Soluble in DMSO or 100% ethanol.
Formel:
C₂₀H₁₈O₄ Lagertemperatur: Gefrierschrank |
MDL:
MFCD00800660 CAS-Nummer: 41060-15-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...