Microcystis aeruginosa Microcystin LY
Lieferant: ENZO LIFE SCIENCES
ALX350148C025
ALX350148C100
ENZOALX350148C025EA
290
EUR
ENZOALX350148C025
ENZOALX350148C100
Microcystis aeruginosa Microcystin LY
Proteine und Peptide
Analog of microcystin-LR (Prod. No. ALX-350-012) with Tyr substituted in place of Arg.
- Isolated from Microcystis aeruginosa.
Its toxicity is intermediate between the more toxic microcystin-LF and the less toxic microcystin-LR.