Orlistat ≥97%

Lieferant: ENZO LIFE SCIENCES

Synonyme: N-Formyl-L-leucine (1S)-1-[[(2S,3S)-3-Hexyl-4-oxo-2-oxetanyl]methyl]dodecyl Ester, (S)-(S)-1-((2S,3S)-3-Hexyl-4-oxooxetan-2-yl)tridecan-2-yl 2-formamido-4-methylpentanoate

ALX350152M250
ENZOALX350152M250EA 217 EUR
ENZOALX350152M250
Orlistat ≥97%
Orlistat

Fatty acid synthase inhibitor


Cell permeable, irreversible inhibitor of gastric and pancreatic lipases. Shows only minimal activity against amylase, trypsin, chymotrypsin, or phospholipase A2 (PLA2). Partially inhibits the hydrolysis of triglycerides and lowers the absorption of dietary fat and promotes weight loss. Anti-obesity drug. Exhibits antitumor activity by inhibition of the thioesterase domain of fatty acid synthase (FAS) both in vitro and in vivo.


Synthetic. Originally isolated from Streptomyces sp.

Formel: C₂₉H₅₃NO₅
Molecular Weight: 495.75 g/mol
MDL: MFCD05662360
CAS-Nummer: 96829-58-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR