R-(-)-Glucoraphanin potassium salt ≥99% (durch HPLC)
Lieferant: ENZO LIFE SCIENCES
Synonyme:
Glucoraphanin, (((5-(Methylsulphinyl)-1-(((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)thio)pentylidene)amino)oxy)sulphonic acid
ALX350181M010
ENZOALX350181M010EA
407
EUR
ENZOALX350181M010
R-(-)-Glucoraphanin potassium salt ≥99% (durch HPLC)
R-(-)-Glucoraphanin potassium salt
Cytochrome P450 modulator
Isolated from Brassica oleracea var. palmifolia££Modulator of hepatic cytochrome P450 and phase II conjugation activity. The enzyme myrosinase transforms R-(-)-glucoraphanin into R-Sulforaphane (Prod. No. ALX-350-230).
Molecular Weight:
475,58 g/mol (anhy) |
MDL:
MFCD08460269 CAS-Nummer: 21414-41-5 |