DL-sulphoraphane (synthetisch) ≥98% (durch UPLC)
Lieferant: ENZO LIFE SCIENCES
Synonyme:
1-Isothiocyanato-4-(methylsulphinyl)butane, 1-Isothiocyanato-4-methylsulphinyl)butane, sulphoraphane
R,S-Sulforaphane is a synthetic version of the naturally occurring compound, R-sulforaphane, from Broccoli. It is a racemic mixture of both the R and S isomers.
Potent, selective inducer of phase II detoxification enzymes. Inhibits chemically induced mammary tumor formation in rats. Inhibits LPS-induced HMGB1 (high mobility group box 1) secretion.
- Carcinogenesis inhibitor
Slightly yellowish liquid.
Soluble in methanol or DMSO.
Formel:
C₆H₁₁NOS₂ Molecular Weight: 177,3 g/mol Lagertemperatur: Gefrierschrank |
MDL:
MFCD00198068 CAS-Nummer: 4478-93-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...