Chlortetracyclin Hydrochlorid

Lieferant: ENZO LIFE SCIENCES
Gefahr

Synonyme: Chlortetracycline HCI, Aureomycin, 7-Chlorotetracycline hydrochloride, (4S,4aS,5aS,6S,12aS)-7-Chloro-4-(dimethylamino)-3,6,10,12,12a-pentahydroxy-6-methyl-1,11-dioxo-1,4,4a,5,5a,6,11,12a-octahydrotetracene-2-carboxamide hydrochloride

ALX350238G005
ENZOALX350238G005EA 72.6 EUR
ENZOALX350238G005
Chlortetracyclin Hydrochlorid
Chlortetracyclin Hydrochlorid
Formel: C₂₂H₂₃ClN₂O₈·HCl
Molecular Weight: 515,34 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD26142699
CAS-Nummer: 64-72-2
EINECS: 200-591-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR