Butein ≥98% (durch HPLC)

Lieferant: ENZO LIFE SCIENCES

Synonyme: 2',3,4,4'-Tetrahydroxychalcone, (E)-1-(2,4-Dihydroxyphenyl)-3-(3,4-dihydroxyphenyl)-2-propen-1-one

ALX350246M010
ENZOALX350246M010EA 355 EUR
ENZOALX350246M010
Butein ≥98% (durch HPLC)
Butein

Tyrosine kinase inhibitor


Plant polyphenol. Specific tyrosine kinase inhibitor. Potently inhibits the tyrosine kinase activity of the EGF receptor and p60c-src. Potent antioxidant and anti-inflammatory agent. Inhibits glutathione reductase and rat liver glutathione S-transferase. Activator of human deacetylase SIRT1. Inhibits aromatase, showing chemopreventive properties. Directly inhibits IKK.

Formel: C₁₅H₁₂O₅
Molecular Weight: 272,26 g/mol
MDL: MFCD00017300
CAS-Nummer: 487-52-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR