Genistin ≥98% (durch HPLC, TLC)

Lieferant: ENZO LIFE SCIENCES

Synonyme: 5-Hydroxy-3-(4-hydroxyphenyl)-7-(((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-4H-chromen-4-one

ALX350247M010
ENZOALX350247M010EA 245 EUR
ENZOALX350247M010
Genistin ≥98% (durch HPLC, TLC)
Genistin

TdT inhibitor


Glucoside of genistein (Prod. No. ALX-350-006) found in soy beans. Useful as a negative control for genistein and other tyrosine kinase inhibitors. Selective inhibitor of terminal deoxyribonucleotidyltransferase (TdT). Displays antioxidant and anticarcinogenic properties.

Formel: C₂₁H₂₀O₁₀
Molecular Weight: 432,38 g/mol
MDL: MFCD00016883
CAS-Nummer: 529-59-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR