Daidzin ≥98%

Lieferant: ENZO LIFE SCIENCES

Synonyme: 7-(beta-D-Glucopyranosyloxy)-4'-hydroxyisoflavone, 3-(4-Hydroxyphenyl)-7-(((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-4H-chromen-4-one

ALX350248M010
ENZOALX350248M010EA 626 EUR
ENZOALX350248M010
Daidzin ≥98%
Daidzin

Glucoside of daidzein.


Glucoside of the isoflavone daidzein found in soy beans.


Soluble in DMSO or dilute aqueous base; insoluble in water.

Formel: C₂₁H₂₀O₉
Molecular Weight: 416.38 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00017466
CAS-Nummer: 552-66-9
Merck Index: 14,02801

Bestellen

Spezifikation Testergebnisse

Purity ≥98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR