(S)-(+)-Abscisic acid cis- und trans-Gemisch (aus Curvularia lunata) ≥97% (durch HPLC)
Lieferant: ENZO LIFE SCIENCES
|
Achtung
|
ALX350255C500
ALX350255M001
ENZOALX350255C500EA
172
EUR
ENZOALX350255C500
ENZOALX350255M001
(S)-(+)-Abscisic acid cis- und trans-Gemisch (aus Curvularia lunata) ≥97% (durch HPLC)
(S)-(+)-Abscisic acid
Plant hormone.
Natural and active isomer of the abscission accelerating plant hormone.
Soluble in ethanol or DMSO.
Formel:
C₁₅H₂₀O₄ Lagertemperatur: Gefrierschrank |
MDL:
MFCD00066545 CAS-Nummer: 21293-29-8 Merck Index: 14,00011 |
Spezifikation Testergebnisse
Purity | ≥97% by HPLC |
Source | Isolated from Curvularia lunata |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...