(S)-(+)-Abscisic acid cis- und trans-Gemisch (aus Curvularia lunata) ≥97% (durch HPLC)

Lieferant: ENZO LIFE SCIENCES
Achtung

ALX350255C500 ALX350255M001
ENZOALX350255C500EA 172 EUR
ENZOALX350255C500 ENZOALX350255M001
(S)-(+)-Abscisic acid cis- und trans-Gemisch (aus Curvularia lunata) ≥97% (durch HPLC)
(S)-(+)-Abscisic acid

Plant hormone.


Natural and active isomer of the abscission accelerating plant hormone.


Soluble in ethanol or DMSO.

Formel: C₁₅H₂₀O₄
Lagertemperatur: Gefrierschrank
MDL: MFCD00066545
CAS-Nummer: 21293-29-8
Merck Index: 14,00011

Bestellen

Spezifikation Testergebnisse

Purity ≥97% by HPLC
Source Isolated from Curvularia lunata


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR