Vinblastine sulphate ≥98%

Lieferant: ENZO LIFE SCIENCES

ALX350257M025
ENZOALX350257M025EA 370 EUR
ENZOALX350257M025
Vinblastine sulphate ≥98%
Vinblastine sulphate

Cell cycle blocker


Plant alkaloid that arrests the cell cycle in G2/M phase by blocking mitotic spindle formation. Inhibits normal microtubule asembly and causes aberant tubulin polymerization. Blocks binding of tubulin to microtubule proteins. Induces apoptosis. Triggers Raf-1 activation, phosphorylation of Bcl-2 family proteins and induction of p53 expression. Cancer chemotherapeutic agent.


Semisynthetic from catharanthine or vindoline.

Formel: C₄₆H₆₀N₄O₁₃S
MDL: MFCD00082457
CAS-Nummer: 143-67-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR