Isoxanthohumol ≥98% (durch 1H-NMR)

Lieferant: ENZO LIFE SCIENCES
Achtung

Synonyme: IXN, 4,7-Dihydroxy-5-methoxy-8-prenylflavanone, 2,3-Dihydro-7-hydroxy-2-(4-hydroxyphenyl)-5-methoxy-8-(3-methyl-2-butenyl)-4H-1-benzopyran-4-one, Humulol, 4',7-Dihydroxy-5-methoxy-8-prenylflavanone

ALX350279M001
ENZOALX350279M001EA 103 EUR
ENZOALX350279M001
Isoxanthohumol ≥98% (durch 1H-NMR)
Isoxanthohumol

Phytoestrogen.


Racemic mixture of (+/-)-Isoxanthohumol. Prenylated flavonoid. Induces apoptosis in mature adipocytes.


Soluble in DMSO (50 mg/ml) or methanol (50 mg/ml).

Formel: C₂₁H₂₂O₅
Molecular Weight: 354,40 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00238718
CAS-Nummer: 70872-29-6
UN: 3077
ADR: 9,III

Bestellen

Spezifikation Testergebnisse

Purity ≥98% (¹H-NMR)
Identity Determined by ¹H-NMR, ¹³C-NMR and LC-MS
Source Synthetic

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR