Xanthohumol ≥98% (durch HPLC)

Lieferant: ENZO LIFE SCIENCES

Synonyme: 2',4,4'-Trihydroxy-6'-methoxy-3'-prenylchalcone, (E)-1-(2,4-Dihydroxy-6-methoxy-3-(3-methylbut-2-en-1-yl)phenyl)-3-(4-hydroxyphenyl)prop-2-en-1-one, (E)-1-[2,4-Dihydroxy-6-methoxy-3-(3-methyl-2-butenyl)phenyl]-3-(4-hydroxyphenyl)propenone, 2',4,4'-Trihydroxy-6'-methoxy-3'-(3-methylbut-2-en-1-yl)chalcone

ALX350280M005
ENZOALX350280M005EA 257 EUR
ENZOALX350280M005
Xanthohumol ≥98% (durch HPLC)
Xanthohumol

DGAT inhibitor


Prenylated chalcone found in beer. Potent inhibitor of diacylglycerol acetyltransferase (DGAT). Inhibits DNA polymerase and induces cell differentiation. Has antiproliferative and cytotoxic effects in human cancer cell lines. Inhibits human P450 enzymes. Induces quinone reductase. Inhibits the expression of HIF-1α and VEGF under hypoxic conditions. Induces apoptosis in mature adipocytes.


Isolated from hops (Humulus lupulus).

Formel: C₂₁H₂₂O₅
MDL: MFCD00210576
CAS-Nummer: 6754-58-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR