Manzamine A ≥98% (durch HPLC)
Lieferant: ENZO LIFE SCIENCES
ALX350294M001
ENZOALX350294M001EA 327 EUR
ENZOALX350294M001
Manzamine A ≥98% (durch HPLC)
Manzamine A
GSK-3 inhibitor
  • Yellow oil
β-Carboline alkaloid with anti-inflammatory, antimalarial, antifungal, anti-HIV-1 and insecticidal activity. Also active against the Gram-positive bacteria Bacillus subtilis and Staphylococcus aureus. Inhibits GSK-3. Has native fluorescence, highest when solubilized in methanol (Ex: 340nm, Em: 387nm).
Isolated from Xestospongia sp.
CAS-Nummer: 104196-68-1