3-O-Acetyl-11-keto-β-boswellic acid (aus Boswellia serrata) ≥98% (durch HPLC)
Lieferant: ENZO LIFE SCIENCES
Shows antitumor and anti-inflammatory activities. Potent non-redox, noncompetitive 5-lipoxygenase and topoisomerase I and IIa inhibitor leading to apoptosis. 10-times more potent than 3-O-acetyl-β-boswellic acid. Exhibits in vivo efficacy in tumor growth and inhibition.
- Inhibitor of lipoxygenase and topoisomerase
White to off-white solid.
Soluble in acetone, dichloromethane, diethyl ether or DMSO.
Formel:
C₃₂H₄₈O₅ Lagertemperatur: Gefrierschrank |
MDL:
MFCD03788777 CAS-Nummer: 67416-61-9 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...