(-)-Arctigenin (aus Arctium lappa) ≥97% (durch HPLC)

Lieferant: ENZO LIFE SCIENCES

Synonyme: Arctigenin, (3R,4R)-4-[(3,4-Dimethoxyphenyl)methyl]dihydro-3-[(4-hydroxy-3-methoxyphenyl)methyl]-2(3H)-furanone, (3R,4R)-4-(3,4-Dimethoxybenzyl)-3-(4-hydroxy-3-methoxybenzyl)dihydrofuran-2(3H)-one

ALX350312M025
ENZOALX350312M025EA 124 EUR
ENZOALX350312M025
(-)-Arctigenin (aus Arctium lappa) ≥97% (durch HPLC)
(-)-Arctigenin

Lignan derivative, which shows antitumor, anti-inflammatory, immunomodulatory and neuroprotective activities. Down-regulates anti-apoptotic protein Bcl-XL. Potent inhibitor of HIV type-I integrase and DNA topoisomerase II. Shows phytoestrogenic and cytotoxic properties against Hep62 cells.  Antagonist for platelet activating factor and Ca²⁺. Blocks the activation of Akt (protein kinase B; PKB) induced by glucose starvation.


  • Antitumor agent


Colorless crystals.


Soluble in DMSO (34mg/ml, heating, sonication); sparingly soluble in water.

Formel: C₂₁H₂₄O₆
Molecular Weight: 372,4 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00870597
CAS-Nummer: 7770-78-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR