(-)-Arctiin (aus Arctium lappa) ≥95% (durch HPLC)

Lieferant: ENZO LIFE SCIENCES

Synonyme: (3R,4R)-4-[(3,4-Dimethoxyphenyl)methyl]-3-[[4-(beta-D-glucopyranosyloxy)-3-methoxyphenyl]methyl]dihydro-2(3H)-furanone, (3R,4R)-4-[(3,4-Dimethoxyphenyl)methyl]-3-[[4-(β-D-glucopyranosyloxy)-3-methoxyphenyl]methyl]dihydro-2(3H)-furanone, (3R,4R)-4-(3,4-Dimethoxybenzyl)-3-(3-methoxy-4-(((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)benzyl)dihydrofuran-2(3H)-one, Arctiin, Arctigenin-4-glucoside

ALX350318M025
ENZOALX350318M025EA 103 EUR
ENZOALX350318M025
(-)-Arctiin (aus Arctium lappa) ≥95% (durch HPLC)
(-)-Arctiin

Lignan derivative with antitumor activities. Down-regulates anti-apoptotic protein Bcl-XL. Potent inhibitor of HIV type-I integrase and DNA topoisomerase II. Shows phytoestrogenic activity and is an antagonist for platelet activating factor and Ca²⁺.


  • Antitumor agent


White to off-white solid.


Soluble in DMSO (5mg/ml); sparingly soluble in water.

Formel: C₂₇H₃₄O₁₁
Molecular Weight: 534,6 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00870630
CAS-Nummer: 20362-31-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR