Harpagoside ≥98% (durch HPLC)

Lieferant: ENZO LIFE SCIENCES
Achtung

Synonyme: (1S,4aS,5R,7S,7aS)-4a,5-Dihydroxy-7-methyl-1-(((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-1,4a,5,6,7,7a-hexahydrocyclopenta[c]pyran-7-yl cinnamate

ALX350333M025
ENZOALX350333M025EA 611 EUR
ENZOALX350333M025
Harpagoside ≥98% (durch HPLC)
Harpagoside

Anti-inflammatory and anti-diabetic.


Suppresses LPS-induced inducible nitric oxide synthase (iNOS; NOS II) and cyclooxygenase-2 (COX-2) expression through inhibition of NF-κB activation.


Soluble in methanol.

Formel: C₂₄H₃₀O₁₁
Molecular Weight: 494.5 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00017415
CAS-Nummer: 19210-12-9

Bestellen

Spezifikation Testergebnisse

Purity ≥98% by HPLC
Source Isolated from Scrophularia ningpoensis Hemsl (Figwort)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR