5,5'-Diallyl-2,2'-dihydroxybiphenyl (Magnolol) ≥98% (durch HPLC)

Lieferant: ENZO LIFE SCIENCES

ALX350352M025
ENZOALX350352M025EA 395 EUR
ENZOALX350352M025
5,5'-Diallyl-2,2'-dihydroxybiphenyl (Magnolol) ≥98% (durch HPLC)
5,5'-Diallyl-2,2'-dihydroxybiphenyl (Magnolol)

Antitumor and anti-angiogenic agent


  • Isolated from Magnolia officinalis


Potent and highly tolerable antitumor and antiangiogenic compound. Anxiolytic, anti-thrombotic and antibacterial. Shows central depressant effects in vivo. Inhibitor of nitric oxide (NO) and TNF-alpha production in LPS-activated macrophages by the suppression of inducible nitric oxide synthase (iNOS; NOS II) expression (IC50=16.8 µm). Isomeric with honokiol (Prod. No. ALX-350-350).

Formel: C₁₈H₁₈O₂
MDL: MFCD00016658
CAS-Nummer: 528-43-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR