Auraptene ≥95% (durch UPLC)
Lieferant: ENZO LIFE SCIENCES
Synonyme:
(E)-7-((3,7-Dimethylocta-2,6-dien-1-yl)oxy)-2H-chromen-2-one, 7-[[(2E)-3,7-Dimethyl-2,6-octadien-1-yl]oxy]-2H-chromen-2-one, 7-Geranyloxycoumarin
ALX350361M025
ENZOALX350361M025EA
120
EUR
ENZOALX350361M025
Auraptene ≥95% (durch UPLC)
Auraptene
Anti-inflammatory and chemopreventive.
Exerts tumor preventive effects through apoptosis. Suppresses cell proliferation and lipid peroxidation. Acts as an agonist of PPARs. Induces phase II drug-metabolising enzymes.
Soluble in methanol or 100% ethanol.
Formel:
C₁₉H₂₂O₃ Molecular Weight: 298,4 g/mol Lagertemperatur: Gefrierschrank |
MDL:
MFCD00075948 CAS-Nummer: 495-02-3 |
Spezifikation Testergebnisse
Purity | ≥95% (UPLC) |
Identity | Determined by NMR |
Source | Isolated from citrus fruit |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...