3,3'-Methylenediindole ≥98% (durch HPLC)

Lieferant: ENZO LIFE SCIENCES
Achtung

Synonyme: DIM, 3,3'-Diindolylmethane

ALX350370M500
ENZOALX350370M500EA 102 EUR
ENZOALX350370M500
3,3'-Methylenediindole ≥98% (durch HPLC)
3,3'-Methylenediindole

Cytochrome P450 activator.


Dimer of indole-3-carbinol. Anticancer and antineoplastic agent. Induces apoptosis in human cancer cells. Activator of cytochrome P450. Partial agonist of aryl hydrocarbon receptor (AhR).


Soluble in DMSO (200 mg/ml); insoluble in water.

Formel: C₁₇H₁₄N₂
Molecular Weight: 246,31 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00195766
CAS-Nummer: 1968-05-4

Bestellen

Spezifikation Testergebnisse

Purity ≥98% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR