Oridonin ≥98% (durch HPLC)
Lieferant: ENZO LIFE SCIENCES
|
Achtung
|
Synonyme:
Isodonol, (1α,6β,7α,14R)-1,5,6,14-Tetrahydroxy-4,4-dimethyl-8-methylenedecahydro-1H-6,11b-(epoxymethano)-6a,9-methanocyclohepta[a]naphthalen-7(8H)-one, Oridonine
ALX350390M050
ENZOALX350390M050EA
250
EUR
ENZOALX350390M050
Oridonin ≥98% (durch HPLC)
Oridonin
Antitumor agent.
Diterpene with potent anti-tumor activity. Induces apoptosis and autophagy.
Soluble in DMSO (20 mg/ml) or PEG 400; insoluble in water.
Formel:
C₂₀H₂₈O₆ Molecular Weight: 364,4 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD00221762 CAS-Nummer: 28957-04-2 |
Spezifikation Testergebnisse
Purity | ≥98% (HPLC) |
Source | Isolated from Rabdosia rubescens |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...