Hydroxytyrosol Acetate
Lieferant: ENZO LIFE SCIENCES
ALX350404M050
ENZOALX350404M050EA
162
EUR
ENZOALX350404M050
Hydroxytyrosol Acetate
Zellkultur-Ergänzungen und -Zusätze
Hydroxytyrosol acetate is an antioxidant comparable to resveratrol. It is converted to hydroxytyrosol through hydrolysis by esterases.
- Reaches full oxygen radical absorbance capacity (ORAC) when converted to free hydoxytyrosol
- ORAC values of hydroxytyrosol are about 50% higher than resveratrol, 10-fold higher than L(+)-ascorbic acid and 4-fold higher than (-)-epicatechin
- Shown to have anti-cancer and anti-viral effects
- Antibacterial at 400 µg/ml against Streptococcus pyogenes, Staphylococcus aureus, Escherichia coli and Klebsiella pneumoniae