Hydroxytyrosol Acetate
Lieferant: ENZO LIFE SCIENCES
ALX350404M050
ENZOALX350404M050EA 162 EUR
ENZOALX350404M050
Hydroxytyrosol Acetate
Zellkultur-Ergänzungen und -Zusätze
Hydroxytyrosol acetate is an antioxidant comparable to resveratrol. It is converted to hydroxytyrosol through hydrolysis by esterases.
  • Reaches full oxygen radical absorbance capacity (ORAC) when converted to free hydoxytyrosol
  • ORAC values of hydroxytyrosol are about 50% higher than resveratrol, 10-fold higher than L(+)-ascorbic acid and 4-fold higher than (-)-epicatechin
  • Shown to have anti-cancer and anti-viral effects
  • Antibacterial at 400 µg/ml against Streptococcus pyogenes, Staphylococcus aureus, Escherichia coli and Klebsiella pneumoniae
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4e44faaecf51d343d96380ac292bbaa693ebbd0e&vid=7f588f0584c66a3af1169d0761b6e009698c36a8&prid=web&clid=SAPDAM