Microcystin-RR
Lieferant: ENZO LIFE SCIENCES
ALX350432C010
ENZOALX350432C010EA 345 EUR
ENZOALX350432C010
Microcystin-RR
Proteine und Peptide
PP2A inhibitor
Isolated from Microcystis aeruginosa.
Microcystin-RR is the Arg-Arg analog of microcystin-LR (Prod. No. ALX-350-431). Hepatotoxic, although found to be up to 10-fold less toxic than microcystin-LR on i.p. injection in mice. Potent inhibitor of protein phosphatase 2A (PP2A). This product can be used as an analytical standard for water testing or for molecular studies when high purity is required.