20-Hydroxyecdysone ≥95% (durch 1H-NMR), weißes Pulver
Lieferant: ENZO LIFE SCIENCES
Ecdysteroid.
Most widely occurring ecdysteroid in both plant and animal species. Controls cell death during metamorphosis of Drosophila melanogaster.
Soluble in water, 100% ethanol, methanol, DMSO and acetic acid.
Formel:
C₂₇H₄₄O₇ Lagertemperatur: Gefrierschrank |
MDL:
MFCD00036740 CAS-Nummer: 5289-74-7 |
Spezifikation Testergebnisse
Purity | ≥95% (¹H-NMR) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...