Enniatin B (aus Fusarium orthoceras var. enniatum) ≥90% (durch HPLC)

Lieferant: ENZO LIFE SCIENCES

ALX380007M001
ENZOALX380007M001EA 225 EUR
ENZOALX380007M001
Enniatin B (aus Fusarium orthoceras var. enniatum) ≥90% (durch HPLC)
Enniatin B

White to off-white crystalline solid


Ionophore antibiotic. Inhibitor of the S. cerevisiae ABC transporter Pdr5p.Inhibitor of acyl-CoA-cholesterol acyltransferase and of phosphodiesterase. Potential anticancer compound.


Soluble in DMSO or other organic solvents, almost insoluble in water


Verpackung: Shipping temperature: Ambient

Formel: C₃₃H₅₇N₃O₉
Lagertemperatur: Gefrierschrank
CAS-Nummer: 917-13-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR