Streptozocin ≥97%
Lieferant: ENZO LIFE SCIENCES
|
Gefahr
|
ALX3800105100
ALX380010G001
ALX380010M100
ENZOALX3800105100EA
136
EUR
ENZOALX3800105100
ENZOALX380010G001
ENZOALX380010M100
Streptozocin ≥97%
Streptozocin
Widely used diabetes inducer in rodents. Inhibition of β-cell O-GlcNAcase by streptozotocin is the mechanism that accounts for its diabetogenic toxicity. N-nitroso-containing antibiotic, acting as a nitric oxide (NO) donor and vasorelaxant. Relaxes phenylephrine-contracted aortic rings at 10µM. Potent methylating agent for DNA.
- O-GlcNAcase inhibitor
White to pale yellow crystalline solid.
Soluble in water (nH2O at 102.8mg/ml) or 100% ethanol (200 proof at 0.92mg/ml).
Formel:
C₈H₁₅N₃O₇ Lagertemperatur: Gefrierschrank |
MDL:
MFCD00006607 CAS-Nummer: 18883-66-4 EINECS: 242-646-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...