Fusidate sodium ≥98% (durch TLC)

Lieferant: ENZO LIFE SCIENCES

Synonyme: Fusidate Sodium, Fusidic acid sodium, Sodium (Z)-2-((3R,4S,5S,8S,9S,10S,11R,13R,14S,16S)-16-acetoxy-3,11-dihydroxy-4,8,10,14-tetramethyldodecahydro-1H-cyclopenta[a]phenanthren-17(2H,10H,14H)-ylidene)-6-methylhept-5-enoate

ALX380011M050
ENZOALX380011M050EA 50.6 EUR
ENZOALX380011M050
Fusidate sodium ≥98% (durch TLC)
Fusidate sodium

Antibiotic which suppresses nitric oxide (NO) toxicity in pancreatic islet cells. Inhibits protein synthesis by inhibition of elongation factor G at the level of the ribosome.


  • Elongation factor G inhibitor


White to off-white solid.


Soluble in water (50mg/ml).

Formel: C₃₁H₄₇NaO₆
Molecular Weight: 538,701 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD09054714
CAS-Nummer: 751-94-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR