Tobramycin
Lieferant: Enzo Life Sciences
|
Gefahr
|
ALX380018G001
ALX380018M250
ENZOALX380018G001EA
392
EUR
ENZOALX380018G001
ENZOALX380018M250
Tobramycin
Tobramycin
Formel:
C₁₈H₃₇N₅O₉ Lagertemperatur: Kühlschrank |
MDL:
MFCD00077885 CAS-Nummer: 32986-56-4 EINECS: 251-322-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...