Neomycinsulfat
Lieferant: ENZO LIFE SCIENCES
Synonyme:
Neomycin Sulfatsalz
ALX380035G025
ENZOALX380035G025EA
521
EUR
ENZOALX380035G025
Neomycinsulfat
Neomycinsulfat
Formel:
C₂₃H₄₆N₆O₁₃·3H₂SO₄ Molecular Weight: 908,87 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00150827 CAS-Nummer: 1405-10-3 EINECS: 215-773-1 Merck Index: 12,06542 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...