Oligomycin C Streptomyces sp. MST-AS5339 95 HPLC_ASSAY_METHOD

Lieferant: ENZO LIFE SCIENCES

ALX380039M001
ENZOALX380039M001EA 612 EUR
ENZOALX380039M001
Oligomycin C Streptomyces sp. MST-AS5339 95 HPLC_ASSAY_METHOD
Oligomycin C

F-ATPase inhibitor.


Minor component of the oligomycin complex. Macrolide antibiotic that inhibits membrane bound mitochondrial ATPase (F1F0), preventing phosphoryl group transfer. Induces apoptosis.


White to off-white powder


Soluble in 100% ethanol, methanol, dimethyl formamide or DMSO


Verpackung: Shipping temperature: Ambient

Formel: C₄₅H₇₄O₁₀
Molecular Weight: 775.1 g/mol
Lagertemperatur: Gefrierschrank
CAS-Nummer: 11052-72-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR