Polymyxin B-Sulfat

Lieferant: ENZO LIFE SCIENCES
Achtung

ALX380040G001 ALX380040G005
ENZOALX380040G001EA 120 EUR
ENZOALX380040G001 ENZOALX380040G005
Polymyxin B-Sulfat
Polymyxin B sulphate

PKC inhibitor


Antibiotic. Inhibitor of protein kinase C (PKC). Breaks the bacterial membrane by incorporating into the phospholipid of the outer membrane and activating phospholipase.


Isolated from Bacillus polymyxa.

Formel: C₅₅H₉₆N₁₆O₁₃·2H₂SO₄
Lagertemperatur: Raumtemperatur
MDL: MFCD00131991
CAS-Nummer: 1405-20-5
EINECS: 215-774-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR