Hygromycin B (aus Streptomyces hygroscopicus) ≥80% (durch HPLC), Flüssigkeit
Lieferant: ENZO LIFE SCIENCES
ALX380059UM01
ENZOALX380059UM01EA
201
EUR
ENZOALX380059UM01
Hygromycin B (aus Streptomyces hygroscopicus) ≥80% (durch HPLC), Flüssigkeit
Hygromycin B
Antibiotic and antiviral.
Antibiotic with high activity. Inhibits growth of prokaryotic and eukaryotic microorganisms and mammalian cells. Inhibits protein synthesis at the translocation step on the 70S ribosome and causes misreading of the mRNA. Penetrates cells that have been permeabilized by virus infection, hence it can act as an effective antiviral agent.
Dark brown solution
Verpackung: Shipping temperature: Ambient
Formel:
C₂₀H₃₇N₃O₁₃ Lagertemperatur: Kühlschrank |
MDL:
MFCD06795479 CAS-Nummer: 31282-04-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...