Fostriecin sodium salt (aus Streptomyces pulveraceous subsp. fostreus) ≥98% (durch HPLC)

Lieferant: ENZO LIFE SCIENCES

Synonyme: (6R)-5,6-Dihydro-6-[(1E,3R,4R,6R,7Z,9Z,11E)-3,6,13-trihydroxy-3-methyl-4-(phosphonooxy)-1,7,9,11-tridecatetraenyl]-2H-pyran-2-one sodium salt, Fostriecin

ALX380065C010
ENZOALX380065C010EA 304 EUR
ENZOALX380065C010
Fostriecin sodium salt (aus Streptomyces pulveraceous subsp. fostreus) ≥98% (durch HPLC)
Fostriecin sodium salt

Antitumor antibiotic. Catalytic inhibitor of topoisomerase II. Strong protein phosphatase 2A (PP2A) inhibitor (IC50=3.2nM). Weak inhibitor for PP1 (IC50=131µM). No apparent effect on PP2B. The binding site for fostriecin on PP2A is different from that of okadaic acid.


Colorless solid


Soluble in water (50 mg/mL), methanol (10 mg/mL) or 100% ethanol (10 mg/mL)


Verpackung: Shipping temperature: Ambient

Formel: C₁₉H₂₆O₉PNa
Lagertemperatur: Gefrierschrank
CAS-Nummer: 87860-39-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR