7-Oxostaurosporine Streptomyces sp. MST-AS5345 98 HPLC_ASSAY_METHOD
Lieferant: ENZO LIFE SCIENCES
Synonyme:
LCM76-L, BMY 41950, RK 1409
ALX380210M001
ENZOALX380210M001EA
630
EUR
ENZOALX380210M001
7-Oxostaurosporine Streptomyces sp. MST-AS5345 98 HPLC_ASSAY_METHOD
7-Oxostaurosporine
Antibiotic. Potent inhibitor of protein kinase C (PKC). Inhibits the cell cycle at the G2 stage.
- Apoptosis inducer. PKC inhibitor.
Yellow solid.
Soluble in DMSO, dimethyl formamide, methanol or 100% ethanol; slightly soluble in water.
Formel:
C₂₈H₂₄N₄O₄ Molecular Weight: 480.5 g/mol Lagertemperatur: Gefrierschrank |
CAS-Nummer:
125035-83-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...