Piericidin A Streptomyces sp. MST-AS5364 95 HPLC_ASSAY_METHOD

Lieferant: ENZO LIFE SCIENCES

Synonyme: 2-((2E,5E,7E,9R,10R,11E)-10-Hydroxy-3,7,9,11-tetramethyltrideca-2,5,7,11-tetraen-1-yl)-5,6-dimethoxy-3-methylpyridin-4-ol, SN 198E, Piericidin A ethanol solution, IT 143D, Shaoguanmycin B, Piericidin A1, MT 1882-I

ALX380235M002
ENZOALX380235M002EA 380 EUR
ENZOALX380235M002
Piericidin A Streptomyces sp. MST-AS5364 95 HPLC_ASSAY_METHOD
Piericidin A

Antibiotic. Specific and potent inhibitor of NADH-ubiquinone oxidoreductase (complex I) that binds to ubiquinone binding site(s). Inhibits both mitochondrial and bacterial NADH ubiquinone oxidoreductase. Anticancer agent that prevents up-regulation of GRP78, and exhibits cytotoxic activity for etoposide resistant cancer cells under glucose-deprived conditions.


  • NADH-ubiquinone oxidoreductase inhibitor


Pale yellow oil.


Soluble in 100% ethanol, methanol, dimethyl formamide or DMSO. Poorly soluble in water.

Formel: C₂₅H₃₇NO₄
Molecular Weight: 415.6 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD01677956
CAS-Nummer: 2738-64-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR