Trinactin ≥95% (durch HPLC), farbloser Film
Lieferant: ENZO LIFE SCIENCES
Synonyme:
Antibiotic AKD 1D, Antibiotic S 3466B
ALX380238M001
ENZOALX380238M001EA
715
EUR
ENZOALX380238M001
Trinactin ≥95% (durch HPLC), farbloser Film
Trinactin
Cation ionophore.
Cation ionophore with high selectivity for ammonium and potassium. Immunosuppressive agent.
Soluble in ethanol, methanol, DMF or DMSO. Limited water solubility.
Formel:
C₄₃H₇₀O₁₂ Lagertemperatur: Gefrierschrank |
CAS-Nummer:
7561-71-9 |
Spezifikation Testergebnisse
Purity | ≥95% (HPLC) |
Source | Isolated from Streptomyces sp. MST-AS5448 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...