Idarubicin hydrochloride ≥98% (durch HPLC)

Lieferant: ENZO LIFE SCIENCES

Synonyme: 4-Demethoxydaunorubicin, DMDR, Idamycin, Idarubicin HCl, (7S,9S)-9-Acetyl-7-(((2R,4S,5S,6S)-4-amino-5-hydroxy-6-methyltetrahydro-2H-pyran-2-yl)oxy)-6,9,11-trihydroxy-7,8,9,10-tetrahydrotetracene-5,12-dione hydrochloride

ALX380260M005
ENZOALX380260M005EA 223 EUR
ENZOALX380260M005
Idarubicin hydrochloride ≥98% (durch HPLC)
Idarubicin hydrochloride

More potent, lipophilic and antineoplastic analog of daunorubicin. DNA-damaging effect includes DNA oxidation and methylation, DNA intercalation, inhibition of DNA synthesis, induction of DNA strand breaks and delay of cell cycle progression. Inhibitor of topoisomerase IIα. Produces endonucleolytic cleavage and is a marker of apoptosis.


  • Topoisomerase IIα inhibitor


Orange crystalline solid.


Soluble in water or methanol.

Formel: C₂₆H₂₇NO₉·HCl
Molecular Weight: 497,5 · 36,5 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00897212
CAS-Nummer: 57852-57-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR