Amikacin disulphate

Lieferant: ENZO LIFE SCIENCES

Synonyme: (S)-4-Amino-N-((1R,2S,3S,4R,5S)-5-amino-2-(((2S,3R,4S,5S,6R)-4-amino-3,5-dihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-4-(((2R,3R,4S,5S,6R)-6-(aminomethyl)-3,4,5-trihydroxytetrahydro-2H-pyran-2-yl)oxy)-3-hydroxycyclohexyl)-2-hydroxybutanamide bis(sulphate)

ALX380266G001
ENZOALX380266G001EA 142 EUR
ENZOALX380266G001
Amikacin disulphate
Amikacin disulphate

White to off-white crystalline powder


  • Aminoglycoside antibiotic
  • Effective against Gram-negative and Gram-positive bacteria
  • Semisynthetic from kanamycin A


Soluble in water (50 mg/mL)


Verpackung: Shipping temperature: Ambient

Formel: C₂₂H₄₃N₅O₁₃·2H₂SO₄
Molecular Weight: 781,77 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00167475
CAS-Nummer: 39831-55-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR