Cycloheximid ≥95% (durch TLC)

Lieferant: ENZO LIFE SCIENCES

Synonyme: Naramycin A, CHX, 4-[(2R)-2-[(1S,3S,5S)-3,5-Dimethyl-2-oxocyclohexyl]-2-hydroxyethyl]piperidin-2,6-dion, Actidion

ALX380269G001 ALX380269G005
ENZOALX380269G001EA 84.7 EUR
ENZOALX380269G001 ENZOALX380269G005
Cycloheximid ≥95% (durch TLC)
Cycloheximid

Protein synthesis inhibitor


Antibiotic. Inhibits protein synthesis in eukaryotes by interfering with the translocation step. Induces apoptosis in tumor cell lines but can also delay or inhibit apoptosis induced by other agents. Widely used for selection of resistant strains of yeast and fungy, controlled inhibiton of protein synthesis for detecton of short-lived proteins and super-induction of protein expression. At 10µg/ml cycloheximide totally inhibits protein synthesis in mouse thymocytes.


Isolated from Streptomyces griseus.

Formel: C₁₅H₂₃NO₄
MDL: MFCD00082346
CAS-Nummer: 66-81-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR