Cefotaxim Natriumsalz ≥95% (durch HPLC)
Lieferant: ENZO LIFE SCIENCES
Inhibits bacterial cell wall synthesis. Effective against Gram-negative and Gram-positive bacteria.
- Cephalosporin antibiotic
- Resistant against β-lactamase
White to light yellow powder
Soluble in water (>50 mg/mL)
Verpackung: Shipping temperature: Ambient
Formel:
C₁₆H₁₇N₅O₇S₂ Lagertemperatur: Gefrierschrank |
MDL:
MFCD00079073 CAS-Nummer: 64485-93-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...