Cefotaxim Natriumsalz ≥95% (durch HPLC)

Lieferant: ENZO LIFE SCIENCES

ALX380271G001 ALX380271M250
ENZOALX380271G001EA 168 EUR
ENZOALX380271G001 ENZOALX380271M250
Cefotaxim Natriumsalz ≥95% (durch HPLC)
Cefotaxime sodium salt

Inhibits bacterial cell wall synthesis. Effective against Gram-negative and Gram-positive bacteria.


  • Cephalosporin antibiotic
  • Resistant against β-lactamase


White to light yellow powder


Soluble in water (>50 mg/mL)


Verpackung: Shipping temperature: Ambient

Formel: C₁₆H₁₇N₅O₇S₂
Lagertemperatur: Gefrierschrank
MDL: MFCD00079073
CAS-Nummer: 64485-93-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR