Colistin sulphate (from Bacillus colistinus)
Lieferant: ENZO LIFE SCIENCES
ALX380272M500
ENZOALX380272M500EA
165
EUR
ENZOALX380272M500
Colistin sulphate (from Bacillus colistinus)
Proteine und Peptide
Polymyxin antibiotic. Binds to lipids on the cell cytoplasmic membrane. Disrupts cell wall integrity. Effective against Gram-negative bacteria.