Erythromycin (aus Streptomyces erythreus) ≥85% (Trockenmasse)

Lieferant: ENZO LIFE SCIENCES

Synonyme: (-)-Erythromycin

ALX380274G005
ENZOALX380274G005EA 86.9 EUR
ENZOALX380274G005
Erythromycin (aus Streptomyces erythreus) ≥85% (Trockenmasse)
Erythromycin

Macrolide antibiotic. Inhibits protein synthesis (elongation) at the transpeptidation step. Binds to the 23S RNA of the 50S ribosomal subunit. Effective against Gram-negative and Gram-positive bacteria.


White to off-white powder


Soluble in water (50 mg/ml), 100% ethanol or methanol


Verpackung: Shipping temperature: Ambient

Formel: C₃₇H₆₇NO₁₃
Lagertemperatur: Gefrierschrank
MDL: MFCD00084654
CAS-Nummer: 114-07-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR